6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

046928-500MG 046928-1G
101877-670EA 246.15 USD
101877-670 101877-672
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid ≥95%
6-(Trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅F₃N₂O₂
Melting Pt: 238…239 °C
MDL Number: MFCD09258865
CAS Number: 1018828-69-7

Order Now

Specification Test Results

Melting point 238-239°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR