2-Bromo-1-(bromomethyl)-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-4-nitrobenzyl bromide
046927-500MG
046927-1G
101877-666EA
320
USD
101877-666
101877-668
2-Bromo-1-(bromomethyl)-4-nitrobenzene ≥95%
2-Bromo-1-(bromomethyl)-4-nitrobenzene
Hazard: Irritant
Formula:
C₇H₅Br₂NO₂ MW: 294.94 g/mol Melting Pt: 73…74 °C |
MDL Number:
MFCD11100213 CAS Number: 940-05-6 |
Specification Test Results
Melting point | 73-74°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...