Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

046926-500MG 046926-1G
101877-662EA 375.38 USD
101877-662 101877-664
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O₂
MW: 258.20 g/mol
Melting Pt: 125-126°C
MDL Number: MFCD11100212
CAS Number: 860457-99-4

Order Now

Specification Test Results

Melting point 125-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR