2,3-Dichloro-4-formylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Dichloropyridine-4-carboxaldehyde, 2,3-Dichloroisonicotinaldehyde
046922-500MG
046922-1G
101877-650EA
250.77
USD
101877-650
101877-652
2,3-Dichloro-4-formylpyridine ≥95%
2,3-Dichloro-4-formylpyridine
Hazard: Irritant
Formula:
C₆H₃Cl₂NO MW: 176.0000 g/mol Melting Pt: 94-96°C |
MDL Number:
MFCD08277282 CAS Number: 884495-41-4 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...