5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine ≥95%

Supplier: Matrix Scientific

046921-500MG 046921-1G
101877-646EA 232.31 USD
101877-646 101877-648
5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine ≥95%
5-(5-Chloro-2-pyridinyl)-1H-1,2,4-triazol-3-amine

Hazard: Irritant

Formula: C₇H₆ClN₅
MW: 195.61 g/mol
Melting Pt: >300 °C
MDL Number: MFCD11100211
CAS Number: 168893-69-4

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR