Benzyl-4-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(5-Chloro-1,6-dihydro-6-oxopyridazin-4-yl)piperazine, N1-CBZ protected
046919-500MG
046919-1G
046919-5G
101877-636EA
181.54
USD
101877-636
101877-638
101877-640
Benzyl-4-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Benzyl-4-(5-chloro-6-oxo-1,6-dihydro-4-pyridazinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₁₇ClN₄O₃ MW: 348.79 g/mol Melting Pt: 169…170 °C |
MDL Number:
MFCD09864817 CAS Number: 952182-38-6 |
Specification Test Results
Melting point | 169-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...