6-(1,4-Diazepan-1-yl)-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine ≥95%

Supplier: Matrix Scientific

046918-500MG 046918-1G 046918-5G
101877-630EA 747.69 USD
101877-630 101877-632 101877-634
6-(1,4-Diazepan-1-yl)-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine ≥95%
6-(1,4-Diazepan-1-yl)-1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₁₄H₁₈F₃N₅
MDL Number: MFCD09607955
CAS Number: 952182-36-4

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR