Ethyl-8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

046911-500MG 046911-1G 046911-5G
101877-594EA 556.92 USD
101877-594 101877-596 101877-598
Ethyl-8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈BrClN₂O₂
Melting Pt: 155-157°C
MDL Number: MFCD09607953
CAS Number: 951884-22-3

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR