N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulfonamide ≥95%

Supplier: Matrix Scientific

046909-500MG 046909-1G
101877-584EA 556.92 USD
101877-584 101877-586
N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulfonamide ≥95%
N-Methyl-4-{[5-(trifluoromethyl)-2-pyridinyl]oxy}benzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O₃S
MW: 332.3 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD09817448
CAS Number: 952183-74-3

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR