N-Methyl-2-nitrobenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046907-1G
046907-5G
101877-576EA
121.54
USD
101877-576
101877-578
N-Methyl-2-nitrobenzenesulfonamide ≥95%
N-Methyl-2-nitrobenzenesulphonamide
Hazard: Irritant
Formula:
C₇H₈N₂O₄S MW: 216.2100 g/mol Melting Pt: 109…111 °C |
MDL Number:
MFCD01115621 CAS Number: 23530-40-7 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...