1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-2-[(5-chloropyridin-2-yl)amino]-1,3-thiazole

046903-500MG 046903-1G 046903-5G
101877-552EA 556.92 USD
101877-552 101877-554 101877-556
1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone ≥95%
1-{2-[(5-Chloro-2-pyridinyl)amino]-1,3-thiazol-5-yl}-1-ethanone

Hazard: Irritant

Formula: C₁₀H₈ClN₃OS
MW: 253.71 g/mol
Melting Pt: 250…252 °C
MDL Number: MFCD09817445
CAS Number: 952183-72-1

Order Now

Specification Test Results

Melting point 250-252°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR