1-[2-(4-Fluoroanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Acetyl-2-[(4-fluorophenyl)amino]-1,3-thiazole
046902-500MG
046902-1G
101877-546EA
453.85
USD
101877-546
101877-548
1-[2-(4-Fluoroanilino)-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(4-Fluoroanilino)-1,3-thiazol-5-yl]-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₉N₂OFS Melting Pt: 217-219°C |
MDL Number:
MFCD09817444 CAS Number: 952183-71-0 |
Specification Test Results
Melting point | 217-219°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...