1-{2-[(5-Chloro-2-pyridinyl)amino]-4-methyl-1,3-thiazol-5-yl}-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-2-[(5-chloropyridin-2-yl)amino]-4-methyl-1,3-thiazole

046901-500MG 046901-1G 046901-5G
101877-540EA 375.38 USD
101877-540 101877-542 101877-544
1-{2-[(5-Chloro-2-pyridinyl)amino]-4-methyl-1,3-thiazol-5-yl}-1-ethanone ≥95%
1-{2-[(5-Chloro-2-pyridinyl)amino]-4-methyl-1,3-thiazol-5-yl}-1-ethanone

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃OS
MW: 267.74 g/mol
Melting Pt: 262…264 °C
MDL Number: MFCD09817443
CAS Number: 952183-70-9

Order Now

Specification Test Results

Melting point 262-264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR