1-[2-(4-Fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-2-[(4-fluorophenyl)amino]-4-methyl-1,3-thiazole

046900-500MG 046900-1G 046900-5G
101877-534EA 375.38 USD
101877-534 101877-536 101877-538
1-[2-(4-Fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(4-Fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₁N₂OFS
Melting Pt: 165-167°C
MDL Number: MFCD01804834
CAS Number: 952183-69-6

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR