5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
046899-500MG
046899-1G
101877-530EA
747.69
USD
101877-530
101877-532
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Bromo-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₁₃H₈BrClFNO₂ Melting Pt: 148…150 °C |
MDL Number:
MFCD09817442 CAS Number: 952183-68-5 |
Specification Test Results
Melting point | 148-150°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...