5-Bromo-1-(3,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046898-500MG 046898-1G
101877-526EA 747.69 USD
101877-526 101877-528
5-Bromo-1-(3,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Bromo-1-(3,4-dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈BrCl₂NO₂
Melting Pt: 133…135 °C
MDL Number: MFCD09817441
CAS Number: 952183-67-4

Order Now

Specification Test Results

Melting point 133-135°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR