4-Chloro-N-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
046896-500MG
046896-1G
046896-5G
101877-514EA
181.54
USD
101877-514
101877-516
101877-518
4-Chloro-N-methylbenzenesulfonamide ≥95%
4-Chloro-N-methylbenzenesulfonamide
Hazard: Irritant
Formula:
C₇H₈ClNO₂S Melting Pt: 52…54 °C |
MDL Number:
MFCD00464330 CAS Number: 6333-79-5 |
Specification Test Results
Melting point | 52-54°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...