Methyl-4-(4-chlorophenyl)-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate ≥95%
Supplier: Matrix Scientific
046893-500MG
046893-1G
046893-5G
101877-496EA
181.54
USD
101877-496
101877-498
101877-500
Methyl-4-(4-chlorophenyl)-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate ≥95%
Methyl-4-(4-chlorophenyl)-6-(2-methoxy-2-oxoethyl)-2-oxo-1,2,3,4-tetrahydro-5-pyrimidine carboxylate
Hazard: Irritant
Formula:
C₁₅H₁₅ClN₂O₅ MW: 338.75 g/mol Melting Pt: 164…165 °C |
MDL Number:
MFCD09817438 CAS Number: 952183-64-1 |
Specification Test Results
Melting point | 164-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...