Ethyl-6-(2-ethoxy-2-oxoethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

046892-500MG 046892-1G 046892-5G
101877-490EA 181.54 USD
101877-490 101877-492 101877-494
Ethyl-6-(2-ethoxy-2-oxoethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-6-(2-ethoxy-2-oxoethyl)-2-oxo-4-phenyl-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₅
MW: 332.36 g/mol
Melting Pt: 185…188 °C
MDL Number: MFCD09817437
CAS Number: 938189-69-6

Order Now

Specification Test Results

Melting point 185-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR