Methyl-2,6-dichloroisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,6-dichloropyridine-4-carboxylate, Methyl 2,6-dichloroisonicotinate, 2,6-Dichloro-4-pyridinecarboxylic acid methyl ester

046891-1G 046891-5G
101877-484EA 53.85 USD
101877-484 101877-486
Methyl-2,6-dichloroisonicotinate ≥95%
Methyl 2,6-dichloroisonicotinate

Hazard: Irritant

Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 83-85°C
MDL Number: MFCD00044409
CAS Number: 42521-09-5

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR