N-Allyl-1H-1,2,3-benzotriazole-1-carbothioamide ≥95%
Supplier: Matrix Scientific
046890-500MG
046890-1G
101877-480EA
375.38
USD
101877-480
101877-482
N-Allyl-1H-1,2,3-benzotriazole-1-carbothioamide ≥95%
N-Allyl-1H-1,2,3-benzotriazole-1-carbothioamide
Hazard: Irritant
Formula:
C₁₀H₁₀N₄S Melting Pt: 48…50 °C |
MDL Number:
MFCD05149230 CAS Number: 690634-06-1 |
Specification Test Results
Melting point | 48-50°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...