3-Bromo-6-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-6-nitroindazole

046889-500MG 046889-1G 046889-5G
101877-474EA 30.77 USD
101877-474 101877-476 101877-478
3-Bromo-6-nitro-1H-indazole ≥95%
3-Bromo-6-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MW: 242.0300 g/mol
Melting Pt: 235…236 °C
MDL Number: MFCD06410881
CAS Number: 70315-68-3

Order Now

Specification Test Results

Melting point 235-236°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR