Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-bromo-4-hydroxy-5-nitrobenzoate
046888-1G
046888-5G
101877-470EA
121.54
USD
101877-470
101877-472
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate ≥95%
Methyl-3-bromo-4-hydroxy-5-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₈H₆BrNO₅ MW: 276.05 g/mol Melting Pt: 131…133 °C |
MDL Number:
MFCD08689690 CAS Number: 40258-72-8 |
Specification Test Results
Melting point | 131-133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...