3-Chloro-5-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
046885-500MG
046885-1G
046885-5G
101877-456EA
32.31
USD
101877-456
101877-458
101877-460
3-Chloro-5-nitro-1H-indazole ≥95%
3-Chloro-5-nitro-1H-indazole
Hazard: Irritant
Formula:
C₇H₄ClN₃O₂ MW: 197.5800 g/mol Melting Pt: 218-220°C |
MDL Number:
MFCD00005692 CAS Number: 4812-45-7 |
Specification Test Results
Melting point | 218-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...