Methyl-2-methyl-1,3-benzoxazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
046884-500MG
046884-1G
046884-5G
101877-450EA
323.08
USD
101877-450
101877-452
101877-454
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-2-methyl-1,3-benzoxazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Melting Pt: 66…68 °C |
MDL Number:
MFCD07774245 CAS Number: 136663-21-3 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...