Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-[amino(hydroxyimino)methyl]benzoate

046883-1G 046883-5G
101877-446EA 218.46 USD
101877-446 101877-448
Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate ≥95%
Methyl-4-[amino(hydroxyimino)methyl]benzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MW: 194.19 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD08689689
CAS Number: 65695-05-8

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR