4-Methyl-7-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
046882-500MG
046882-1G
046882-5G
101877-438EA
375.38
USD
101877-438
101877-440
101877-442
4-Methyl-7-nitro-1H-indazole ≥95%
4-Methyl-7-nitro-1H-indazole
Hazard: Irritant
Formula:
C₈H₇N₃O₂ MW: 177.1601 g/mol Melting Pt: 178-180°C |
MDL Number:
MFCD08689688 CAS Number: 104103-06-2 |
Specification Test Results
Melting point | 178-180°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...