Methyl-3-amino-4-hydroxybenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-amino-4-hydroxybenzoate
046881-1G
046881-5G
101877-434EA
90.77
USD
101877-434
101877-436
Methyl-3-amino-4-hydroxybenzoate ≥95%
Methyl-3-amino-4-hydroxybenzoate
Hazard: Irritant
Formula:
C₈H₉NO₃ Melting Pt: 141…143 °C |
MDL Number:
MFCD06659667 CAS Number: 536-25-4 |
Specification Test Results
Melting point | 141-143°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...