Methyl-1,3-benzoxazole-5-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl benzo[d]oxazole-5-carboxylate
046880-500MG
046880-1G
046880-5G
101877-426EA
104.62
USD
101877-426
101877-428
101877-430
Methyl-1,3-benzoxazole-5-carboxylate ≥97%
Methyl-1,3-benzoxazole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₇NO₃ MW: 177.16 g/mol Melting Pt: 110…111 °C |
MDL Number:
MFCD08689687 CAS Number: 924869-17-0 |
Specification Test Results
Melting point | 110-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...