3-{2-[4-(tert-Butoxycarbonyl)piperazino]ethoxy}-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Piperazin-1-ylethoxy)thiophene-2-carboxylic acid, N4-BOC protected

046879-500MG 046879-1G 046879-5G
101877-420EA 747.69 USD
101877-420 101877-422 101877-424
3-{2-[4-(tert-Butoxycarbonyl)piperazino]ethoxy}-2-thiophenecarboxylic acid ≥95%
3-{2-[4-(tert-Butoxycarbonyl)piperazino]ethoxy}-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₄N₂O₅S
MW: 356.44 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD08689686
CAS Number: 924869-16-9

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR