1-{2-[4-(tert-Butoxycarbonyl)piperazino]ethyl}-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(2-Piperazin-1-ylethyl)piperidine-4-carboxylic acid, N4-BOC protected
046874-500MG
046874-1G
046874-5G
101877-396EA
747.69
USD
101877-396
101877-398
101877-400
1-{2-[4-(tert-Butoxycarbonyl)piperazino]ethyl}-4-piperidinecarboxylic acid ≥95%
1-{2-[4-(tert-Butoxycarbonyl)piperazino]ethyl}-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₇H₃₁N₃O₄ MW: 341.45 g/mol Melting Pt: 160…163 °C |
MDL Number:
MFCD06808600 CAS Number: 874831-74-0 |
Specification Test Results
Melting point | 160-163°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...