Ethyl-3-(4,4-dimethyl-2,6-dioxopiperidino)-benzenecarboxylate ≥95%

Supplier: Matrix Scientific

046873-500MG 046873-1G 046873-5G
101877-390EA 278.46 USD
101877-390 101877-392 101877-394
Ethyl-3-(4,4-dimethyl-2,6-dioxopiperidino)-benzenecarboxylate ≥95%
Ethyl-3-(4,4-dimethyl-2,6-dioxopiperidino)-benzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₉NO₄
Melting Pt: 134…136 °C
MDL Number: MFCD09152739
CAS Number: 952183-63-0

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR