3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(5-Bromopyridin-3-yl)-1,3,4-oxadiazole
046870-500MG
046870-1G
101877-376EA
206.15
USD
101877-376
101877-378
3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine ≥95%
3-Bromo-5-(1,3,4-oxadiazol-2-yl)pyridine
Hazard: Irritant
Formula:
C₇H₄BrN₃O MW: 226.04 g/mol Melting Pt: 182…183 °C |
MDL Number:
MFCD08444031 CAS Number: 924869-13-6 |
Specification Test Results
Melting point | 182-183°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...