2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-[(3,4-Dichlorobenzyl)thio]-1-methyl-1H-imidazole-5-carboxaldehyde
046868-500MG
046868-1G
046868-5G
101877-364EA
556.92
USD
101877-364
101877-366
101877-368
2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazole-5-carbaldehyde ≥95%
2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀Cl₂N₂OS MW: 301.2 g/mol Melting Pt: 95…97 °C |
MDL Number:
MFCD08689685 CAS Number: 924869-11-4 |
Specification Test Results
Melting point | 95-97°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...