8H-Indeno[1,2-d][1,3]thiazol-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-8H-indeno[1,2-d][1,3]thiazole

046866-500MG 046866-1G 046866-5G
101877-354EA 278.46 USD
101877-354 101877-356 101877-358
8H-Indeno[1,2-d][1,3]thiazol-2-amine ≥95%
8H-Indeno[1,2-d][1,3]thiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₈N₂S
Melting Pt: 268…270 °C
MDL Number: MFCD08689683
CAS Number: 85787-95-7

Order Now

Specification Test Results

Melting point 268-270°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR