4,5-Dimethyl-1,3-thiazol-2-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
4,5-Dimethylthiazol-2-amine, 2-Amino-4,5-dimethyl-1,3-thiazole
046865-500MG
046865-1G
101877-350EA
38.46
USD
101877-350
101877-352
4,5-Dimethyl-1,3-thiazol-2-amine ≥95%
4,5-Dimethyl-1,3-thiazol-2-amine
Hazard: Irritant
Formula:
C₅H₈N₂S MW: 128.2 g/mol Melting Pt: 66…69 °C |
MDL Number:
MFCD00462425 CAS Number: 2289-75-0 |
Specification Test Results
Melting point | 66-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...