5-Amino-2-chloro-4-nitrobenzotrifluoride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Nitro-4-chloro-5-(trifluoromethyl)aniline, 5-Amino-2-chloro-4-nitrobenzotrifluoride
046864-500MG
046864-1G
046864-5G
101877-344EA
278.46
USD
101877-344
101877-346
101877-348
5-Amino-2-chloro-4-nitrobenzotrifluoride ≥95%
5-Amino-2-chloro-4-nitrobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₄ClF₃N₂O₂ Melting Pt: 123-125°C |
MDL Number:
MFCD03094264 CAS Number: 167415-22-7 |
Specification Test Results
Melting point | 123-125°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...