Methyl-5-(4-methoxyphenyl)-1-benzothiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

046862-500MG 046862-1G
101877-334EA 556.92 USD
101877-334 101877-336
Methyl-5-(4-methoxyphenyl)-1-benzothiophene-2-carboxylate ≥95%
Methyl-5-(4-methoxyphenyl)-1-benzothiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₄O₃S
MW: 298.36 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD08689681
CAS Number: 924869-09-0

Order Now

Specification Test Results

Melting point 185-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR