N-[4-Chloro-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-nitro-N-(trifluoroacetyl)-5-(trifluoromethyl)aniline

046861-500MG 046861-1G 046861-5G
101877-328EA 181.54 USD
101877-328 101877-330 101877-332
N-[4-Chloro-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide ≥95%
N-[4-Chloro-2-nitro-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₉H₃ClF₆N₂O₃
Melting Pt: 100…102 °C
MDL Number: MFCD08689680
CAS Number: 167415-21-6

Order Now

Specification Test Results

Melting point 100-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR