2-(Acetylamino)-3-[(4-chlorophenyl)sulfanyl]propanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Acetylamino)-3-[(4-chlorophenyl)thio]propanoic acid
046859-500MG
046859-1G
046859-5G
101877-316EA
201.54
USD
101877-316
101877-318
101877-320
2-(Acetylamino)-3-[(4-chlorophenyl)sulfanyl]propanoic acid ≥95%
2-(Acetylamino)-3-[(4-chlorophenyl)sulfanyl]propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃S MW: 273.74 g/mol Melting Pt: 144…147 °C |
MDL Number:
MFCD09027090 CAS Number: 21056-72-4 |
Specification Test Results
Melting point | 144-147°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...