Ethyl-4-ethoxy-2-phenyl-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
046858-500MG
046858-1G
046858-5G
101877-310EA
556.92
USD
101877-310
101877-312
101877-314
Ethyl-4-ethoxy-2-phenyl-5-pyrimidinecarboxylate ≥95%
Ethyl-4-ethoxy-2-phenyl-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O₃ MW: 272.31 g/mol Melting Pt: 54…56 °C |
MDL Number:
MFCD00483313 CAS Number: 122773-99-3 |
Specification Test Results
Melting point | 54-56°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...