{1-[(4-Chlorophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(4-Chlorophenyl)sulphonyl]-3-(hydroxymethyl)piperidine

046857-500MG 046857-1G 046857-5G
101877-304EA 181.54 USD
101877-304 101877-306 101877-308
{1-[(4-Chlorophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%
{1-[(4-Chlorophenyl)sulfonyl]-3-piperidinyl}methanol

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₃S
MW: 289.78 g/mol
Melting Pt: 117…118 °C
MDL Number: MFCD01185759
CAS Number: 349098-57-3

Order Now

Specification Test Results

Melting point 117-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR