Ethyl-4-[2-(3-chlorophenyl)-2-hydroxyethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-[2-(3-chlorophenyl)-2-hydroxyethyl]piperazine-1-carboxylate

046856-500MG 046856-1G 046856-5G
101877-298EA 278.46 USD
101877-298 101877-300 101877-302
Ethyl-4-[2-(3-chlorophenyl)-2-hydroxyethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-[2-(3-chlorophenyl)-2-hydroxyethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O₃
MW: 312.8 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD08689678
CAS Number: 486439-08-1

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR