Methyl-2-amino-2-[4-(trifluoromethyl)phenyl]acetate hydrochloride ≥95%
Supplier: Matrix Scientific
046855-500MG
046855-1G
101877-294EA
376.92
USD
101877-294
101877-296
Methyl-2-amino-2-[4-(trifluoromethyl)phenyl]acetate hydrochloride ≥95%
Methyl-2-amino-2-[4-(trifluoromethyl)phenyl]acetate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₁ClF₃NO₂ Melting Pt: 176…179 °C |
MDL Number:
MFCD08689677 CAS Number: 390815-48-2 |
Specification Test Results
Melting point | 176-179°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...