Ethyl-4-[2-(3-chlorophenyl)-2-oxoethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
046854-500MG
046854-1G
046854-5G
101877-288EA
181.54
USD
101877-288
101877-290
101877-292
Ethyl-4-[2-(3-chlorophenyl)-2-oxoethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Ethyl-4-[2-(3-chlorophenyl)-2-oxoethyl]tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₉ClN₂O₃ Melting Pt: 80…82 °C |
MDL Number:
MFCD08444029 CAS Number: 486439-06-9 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...