{1-[(4-Bromophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(4-Bromophenyl)sulphonyl]-3-(hydroxymethyl)piperidine

046849-500MG 046849-1G 046849-5G
101877-260EA 181.54 USD
101877-260 101877-262 101877-264
{1-[(4-Bromophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%
{1-[(4-Bromophenyl)sulfonyl]-3-piperidinyl}methanol

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₃S
MW: 334.24 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD08444025
CAS Number: 924869-08-9

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR