{1-[(4-Nitrophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Hydroxymethyl)-1-[(4-nitrophenyl)sulphonyl]piperidine
046848-500MG
046848-1G
046848-5G
101877-254EA
278.46
USD
101877-254
101877-256
101877-258
{1-[(4-Nitrophenyl)sulfonyl]-3-piperidinyl}methanol ≥95%
{1-[(4-Nitrophenyl)sulfonyl]-3-piperidinyl}methanol
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₅S MW: 300.34 g/mol Melting Pt: 135…138 °C |
MDL Number:
MFCD01212730 CAS Number: 349098-92-6 |
Specification Test Results
Melting point | 135-138°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...