Ethyl-2-(3-chloro-4-fluoroanilino)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl [(3-chloro-4-fluorophenyl)amino]acetate

046847-500MG 046847-1G 046847-5G
101877-248EA 129.23 USD
101877-248 101877-250 101877-252
Ethyl-2-(3-chloro-4-fluoroanilino)acetate ≥95%
Ethyl-2-(3-chloro-4-fluoroanilino)acetate

Hazard: Irritant

Formula: C₁₀H₁₁ClFNO₂
Melting Pt: 108…110 °C
MDL Number: MFCD08689674
CAS Number: 2344-98-1

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR