3-(1,3-Benzoxazol-2-yl)-2-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(1,3-Benzoxazol-2-yl)-2-hydroxypyridine

046843-500MG 046843-1G 046843-5G
101877-224EA 181.54 USD
101877-224 101877-226 101877-228
3-(1,3-Benzoxazol-2-yl)-2-pyridinol ≥95%
3-(1,3-Benzoxazol-2-yl)-2-pyridinol

Hazard: Irritant

Formula: C₁₂H₈N₂O₂
MW: 212.21 g/mol
Melting Pt: >300 °C
MDL Number: MFCD08444020
CAS Number: 292140-74-0

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR