(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione ≥95%

Supplier: Matrix Scientific

046841-500MG 046841-1G
101877-214EA 415.38 USD
101877-214 101877-216
(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione ≥95%
(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂
Melting Pt: 157…158 °C
MDL Number: MFCD04116221
CAS Number: 370879-53-1

Order Now

Specification Test Results

Melting point 157-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR